6-Methyl-2-oxo-4-thioxo-1,2,3,4-tetrahydro-pyrimidine-5-carboxylic acid ethyl ester ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-methyl-2-oxo-1,2,3,4-tetrahydro-4-thioxopyrimidine-5-carboxylate

083503-1G 083503-10G
102839-952EA 149.23 USD
102839-952 102839-954
6-Methyl-2-oxo-4-thioxo-1,2,3,4-tetrahydro-pyrimidine-5-carboxylic acid ethyl ester ≥97%
6-Methyl-2-oxo-4-thioxo-1,2,3,4-tetrahydro-pyrimidine-5-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₈H₁₀N₂O₃S
MW: 214.24 g/mol
MDL Number: MFCD06411556
CAS Number: 13996-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR