Methyl-5-methylpyrazine-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 5-methylpyrazine-2-carboxylate

083498-1G 083498-10G 083498-50G
102839-934EA 156.92 USD
102839-934 102839-936 102839-938
Methyl-5-methylpyrazine-2-carboxylate ≥97%
Methyl-5-methylpyrazine-2-carboxylate

Hazard: Irritant

Formula: C₇H₈N₂O₂
MW: 152.1500 g/mol
MDL Number: MFCD04116910
CAS Number: 41110-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR