Diethyl-2-propyl-1H-imidazole-4,5-dicarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Propyl-1H-imidazole-4,5-dicarboxylic acid diethyl ester, Diethyl 2-propyl-1H-imidazole-4,5-dicarboxylate

083496-1G 083496-10G
102839-928EA 15.38 USD
102839-928 102839-930
Diethyl-2-propyl-1H-imidazole-4,5-dicarboxylate ≥97%
Diethyl-2-propyl-1H-imidazole-4,5-dicarboxylate

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₄
MW: 254.2800 g/mol
MDL Number: MFCD08062365
CAS Number: 144689-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR