Ethyl-3-(2-thienyl)-1H-pyrazole-5-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-(2-Thienyl)pyrazole-3-carboxylate

083494-1G 083494-10G
102839-922EA 200 USD
102839-922 102839-924
Ethyl-3-(2-thienyl)-1H-pyrazole-5-carboxylate ≥97%
Ethyl-3-(2-thienyl)-1H-pyrazole-5-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂S
MW: 222.262 g/mol
MDL Number: MFCD00099767
CAS Number: 121195-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR