Methyl-5-methylisoxazole-3-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 5-methylisoxazole-3-carboxylate
083479-1G
083479-10G
083479-50G
102839-886EA
72.31
USD
102839-886
102839-888
102839-890
Methyl-5-methylisoxazole-3-carboxylate ≥97%
Methyl 5-methylisoxazole-3-carboxylate
Hazard: Irritant
Formula:
C₆H₇NO₃ MW: 141.13 g/mol |
MDL Number:
MFCD00015895 CAS Number: 19788-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...