Methyl-3-(2,6-dichlorophenyl)-5-methylisoxazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

083478-1G 083478-10G
102839-882EA 127.69 USD
102839-882 102839-884
Methyl-3-(2,6-dichlorophenyl)-5-methylisoxazole-4-carboxylate ≥97%
Methyl-3-(2,6-dichlorophenyl)-5-methylisoxazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₉Cl₂NO₃
MW: 286.12 g/mol
MDL Number: MFCD00115005
CAS Number: 4402-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR