Methyl-3-methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

083476-1G 083476-10G
102839-876EA 383.08 USD
102839-876 102839-878
Methyl-3-methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylate ≥97%
Methyl-3-methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₄
MDL Number: MFCD00085130
CAS Number: 175277-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR