Methyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate ≥97%
Supplier: Matrix Scientific
083463-1G
083463-10G
102839-846EA
40
USD
102839-846
102839-848
Methyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate ≥97%
Methyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate
Hazard: Irritant
Formula:
C₆H₁₁NO₃ |
MDL Number:
MFCD00797543 CAS Number: 1499-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...