Ethyl-4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate ≥97%

Supplier: Matrix Scientific

083458-1G 083458-10G
102839-834EA 149.23 USD
102839-834 102839-836
Ethyl-4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate ≥97%
Ethyl-4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₉H₁₂BrNO₂
MW: 246.11 g/mol
Melting Pt: 166 °C
MDL Number: MFCD02647051
CAS Number: 5408-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR