Methyl-3-methyl-2-furoate 97
Supplier: Matrix Scientific
Synonyms:
Methyl 3-methylfuran-2-carboxylate
083451-1G
083451-10G
102839-818EA
127.69
USD
102839-818
102839-820
Methyl-3-methyl-2-furoate 97
Methyl-3-methyl-2-furoate
Hazard: Irritant
Formula:
C₇H₈O₃ MW: 140.14 g/mol |
MDL Number:
MFCD00014110 CAS Number: 6141-57-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...