Ethyl-2-amino-4-(2-thienyl)thiophene-3-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 5'-amino-2,3'-bithiophene-4'-carboxylate

083438-1G 083438-10G
102839-782EA 241.54 USD
102839-782 102839-784
Ethyl-2-amino-4-(2-thienyl)thiophene-3-carboxylate ≥97%
Ethyl-2-amino-4-(2-thienyl)thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₂S₂
MW: 253.34 g/mol
MDL Number: MFCD00115120
CAS Number: 243669-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR