Methyl-3-amino-5-phenylthiophene-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-5-phenylthiophene-2-carboxylate

083433-1G 083433-10G
102839-766EA 56.92 USD
102839-766 102839-768
Methyl-3-amino-5-phenylthiophene-2-carboxylate ≥97%
Methyl-3-amino-5-phenylthiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₂S
MW: 233.2862 g/mol
MDL Number: MFCD00068161
CAS Number: 100063-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR