Methyl-3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate

083432-1G 083432-10G
102839-762EA 143.08 USD
102839-762 102839-764
Methyl-3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate ≥97%
Methyl-3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₂S
MW: 267.74 g/mol
MDL Number: MFCD00052591
CAS Number: 91076-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR