Methyl-2,6-dichloro-5-fluoronicotinate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2,6-dichloro-5-fluoronicotinate

083415-1G 083415-10G
102839-718EA 213.85 USD
102839-718 102839-720
Methyl-2,6-dichloro-5-fluoronicotinate ≥97%
Methyl-2,6-dichloro-5-fluoronicotinate

Hazard: Irritant

Formula: C₇H₄Cl₂FNO₂
MW: 224.02 g/mol
MDL Number: MFCD02180634
CAS Number: 189281-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR