Isopropyl-3-acetylpyridine-2-carboxylate ≥97%

Supplier: Matrix Scientific

083405-1G 083405-10G
102839-696EA 72.31 USD
102839-696 102839-698
Isopropyl-3-acetylpyridine-2-carboxylate ≥97%
Isopropyl-3-acetylpyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD01314548
CAS Number: 195812-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR