Arecolinium bromide ≥97%
Supplier: Matrix Scientific
Synonyms:
Arecoline hydrobromide
083401-1G
083401-10G
083401-50G
102839-682EA
101.54
USD
102839-682
102839-684
102839-686
Arecolinium bromide ≥97%
Arecolinium bromide
Hazard: Irritant
Formula:
C₈H₁₄BrNO₂ |
MDL Number:
MFCD00039041 CAS Number: 300-08-3 UN: 1544 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...