Diethyl-2,6-pyridinedicarboxylate ≥97%

Supplier: Matrix Scientific

083393-1G 083393-10G 083393-50G
102839-654EA 156.92 USD
102839-654 102839-656 102839-658
Diethyl-2,6-pyridinedicarboxylate ≥97%
Diethyl-2,6-pyridinedicarboxylate

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD00033792
CAS Number: 15658-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR