Ethyl-2-methyl-2-phenylpropanoate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl dimethylbenzeneacetate, Ethyl 2-methyl-2-phenylpropanoate
083368-1G
083368-10G
102839-586EA
298.46
USD
102839-586
102839-588
Ethyl-2-methyl-2-phenylpropanoate ≥97%
Ethyl-2-methyl-2-phenylpropanoate
Hazard: Irritant
Formula:
C₁₂H₁₆O₂ MW: 192.26 g/mol |
MDL Number:
MFCD03844687 CAS Number: 2901-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...