Methyl-2-amino-5-fluorobenzoate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-Amino-5-fluorobenzoate, 5-Fluoroanthranilic acid methyll ester, Methyl 5-Fluoroanthranilate, 2-Amino-5-fluorobenzoic acid methyll ester, Methyl 2-amino-5-fluorobenzoate
083365-1G
083365-10G
083365-50G
102839-572EA
15.38
USD
102839-572
102839-574
102839-576
Methyl-2-amino-5-fluorobenzoate ≥97%
Methyl-2-amino-5-fluorobenzoate
Hazard: Irritant
Formula:
C₈H₈FNO₂ MW: 169.16 g/mol |
MDL Number:
MFCD03791308 CAS Number: 319-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...