Ethyl-4-amino-3,5-diiodobenzoate ≥97%

Supplier: Matrix Scientific

083356-1G 083356-10G
102839-548EA 126.15 USD
102839-548 102839-550
Ethyl-4-amino-3,5-diiodobenzoate ≥97%
Ethyl 4-amino-3,5-diiodobenzoate

Hazard: Irritant/Stored under Argon

Formula: C₉H₉I₂NO₂
MW: 416.982 g/mol
MDL Number: MFCD00017092
CAS Number: 5400-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR