Ethyl [4-(benzyloxy)phenoxy]acetate ≥97%

Supplier: Matrix Scientific

083354-1G 083354-10G
102839-540EA 184.62 USD
102839-540 102839-542
Ethyl [4-(benzyloxy)phenoxy]acetate ≥97%
Ethyl [4-(benzyloxy)phenoxy]acetate

Hazard: Irritant

Formula: C₁₇H₁₈O₄
MDL Number: MFCD03847881
CAS Number: 142717-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR