Methyl-3,4,5-trimethoxycinnamate ≥98%

Supplier: Matrix Scientific

083346-1G 083346-10G
102839-516EA 72.31 USD
102839-516 102839-518
Methyl-3,4,5-trimethoxycinnamate ≥98%
Methyl-3,4,5-trimethoxycinnamate

Hazard: Irritant

Formula: C₁₃H₁₆O₅
MW: 252.27 g/mol
MDL Number: MFCD03614740
CAS Number: 20329-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR