Ethyl (4-iodophenyl)acetate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(4-iodophenyl)acetate

083336-1G 083336-10G
102839-490EA 353.85 USD
102839-490 102839-492
Ethyl (4-iodophenyl)acetate ≥97%
Ethyl (4-iodophenyl)acetate

Hazard: Irritant

Formula: C₁₀H₁₁IO₂
MW: 290.0976 g/mol
MDL Number: MFCD07779004
CAS Number: 15250-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR