Methyl-3,4-dimethylbenzoate ≥97%

Supplier: Matrix Scientific

083332-1G 083332-10G 083332-50G
102839-478EA 101.54 USD
102839-478 102839-480 102839-482
Methyl-3,4-dimethylbenzoate ≥97%
Methyl 3,4-dimethylbenzoate

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MDL Number: MFCD00219861
CAS Number: 38404-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR