Methyl-5-acetylsalicylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 5-acetyl-2-hydroxybenzoate

083326-1G 083326-10G 083326-50G
102839-462EA 72.31 USD
102839-462 102839-464 102839-466
Methyl-5-acetylsalicylate ≥97%
Methyl 5-acetylsalicylate

Hazard: Irritant

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
MDL Number: MFCD00040924
CAS Number: 16475-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR