Ethyl-3,5-dichloro-4-hydroxybenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 3,5-dichloro-4-hydroxybenzoate

083325-1G 083325-10G 083325-50G
102839-456EA 44.62 USD
102839-456 102839-458 102839-460
Ethyl-3,5-dichloro-4-hydroxybenzoate ≥97%
Ethyl-3,5-dichloro-4-hydroxybenzoate

Hazard: Irritant

Formula: C₉H₈Cl₂O₃
MDL Number: MFCD00016420
CAS Number: 17302-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR