Methyl-3-formyl-4-hydroxybenzoate ≥97%
Supplier: Matrix Scientific
083277-1G
083277-10G
102839-348EA
153.85
USD
102839-348
102839-350
Methyl-3-formyl-4-hydroxybenzoate ≥97%
Methyl-3-formyl-4-hydroxybenzoate
Hazard: Irritant
Formula:
C₉H₈O₄ Melting Pt: 80…84 °C |
MDL Number:
MFCD01661473 CAS Number: 24589-99-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...