(3,4-Dichlorophenyl)acetic acid ethyl ester ≥97%

Supplier: Matrix Scientific

083252-10G 083252-50G
102839-290EA 258.46 USD
102839-290 102839-292
(3,4-Dichlorophenyl)acetic acid ethyl ester ≥97%
(3,4-Dichlorophenyl)acetic acid ethyl ester

Hazard: Irritant

Formula: (C₆H₃Cl₂)CH₂COOCH₂CH₃
MDL Number: MFCD00833397
CAS Number: 6725-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR