Diethyl (2-phenylethyl)malonate ≥97%

Supplier: Matrix Scientific

Synonyms: Diethyl 2-phenethylmalonate

083248-1G 083248-10G
102839-278EA 80 USD
102839-278 102839-280
Diethyl (2-phenylethyl)malonate ≥97%
Diethyl (2-phenylethyl)malonate

Hazard: Irritant

Formula: C₁₅H₂₀O₄
MW: 264.32 g/mol
Boiling Pt: 60-80°C/0.04 Torr
Density: 1.058
MDL Number: MFCD00026887
CAS Number: 6628-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR