Methyl-ɑ-bromophenylacetate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl alpha-broMophenylacetate, ɑ-Bromophenylacetic Acid Methyl Ester

083244-1G 083244-10G
102839-268EA 72.31 USD
102839-268 102839-270
Methyl-ɑ-bromophenylacetate ≥97%
Methyl-alpha-bromophenylacetate

Hazard: Irritant

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD00013535
CAS Number: 3042-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR