Methyl-4-iodo-2-methoxybenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Iodo-2-(methoxycarbonyl)anisole, Methyl 4-iodo-2-methoxybenzoate, 4-Iodo-2-methoxybenzoic acid methyl ester, Methyl-4-iodo-2-methoxybenzoate, Methyl 4-iodo-o-anisate, Methyl 3-iodo-4-methoxybenzoate

083241-1G 083241-10G
102839-262EA 127.69 USD
102839-262 102839-264
Methyl-4-iodo-2-methoxybenzoate ≥97%
Methyl-4-iodo-2-methoxybenzoate

Hazard: Irritant

Formula: C₉H₉IO₃
MW: 292.07
MDL Number: MFCD00052925
CAS Number: 35387-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR