Methyl-2-(acetylamino)-5-nitrobenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-acetamido-5-nitrobenzoate

083236-1G 083236-10G
102839-246EA 241.54 USD
102839-246 102839-248
Methyl-2-(acetylamino)-5-nitrobenzoate ≥97%
Methyl-2-(acetylamino)-5-nitrobenzoate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₅
MDL Number: MFCD00157624
CAS Number: 5409-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR