Methyl-2,6-dihydroxybenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2,6-dihydroxybenzoate, 2,6-Dihydroxybenzoic acid methyll ester, gamma-Resorcylic acid methyll ester

083234-1G 083234-10G 083234-50G
102839-240EA 306.15 USD
102839-240 102839-242 102839-244
Methyl-2,6-dihydroxybenzoate ≥97%
Methyl-2,6-dihydroxybenzoate

Hazard: Irritant

Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00059619
CAS Number: 2150-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR