2,4-Dibromobutanoyl chloride ≥95%

Supplier: Matrix Scientific

083228-1G 083228-10G
102839-218EA 241.54 USD
102839-218 102839-220
2,4-Dibromobutanoyl chloride ≥95%
2,4-Dibromobutanoyl chloride

Hazard: Irritant

Formula: C₄H₅Br₂ClO
MW: 264.35 g/mol
MDL Number: MFCD01862994
CAS Number: 82820-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR