Benzo[b]thiophene-2-carbonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: Benzo[b]thiophene-2-carbonyl chloride

083183-1G 083183-10G
102839-122EA 120 USD
102839-122 102839-124
Benzo[b]thiophene-2-carbonyl chloride ≥97%
Benzo[b]thiophene-2-carbonyl chlorid

Hazard: Corrosive/Stored under Argon/Moisture Sensitive

Formula: C₉H₅ClOS
MW: 196.65 g/mol
Melting Pt: 85…88 °C
MDL Number: MFCD00051635
CAS Number: 39827-11-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR