3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carbonyl chloride ≥97%

Supplier: Matrix Scientific

083174-1G 083174-10G
102839-100EA 467.69 USD
102839-100 102839-102
3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carbonyl chloride ≥97%
3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carbonyl chloride

Hazard: Irritant

Formula: C₁₂H₁₁ClN₂O
MDL Number: MFCD03421620
CAS Number: 61226-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR