3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonyl chloride ≥95%

Supplier: Matrix Scientific

083166-1G 083166-10G
102839-082EA 86.15 USD
102839-082 102839-084
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonyl chloride ≥95%
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonyl chloride

Hazard: Irritant

Formula: C₁₁H₆Cl₂FNO₂
MW: 274.08 g/mol
MDL Number: MFCD00055650
CAS Number: 69399-79-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR