3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride ≥95%

Supplier: Matrix Scientific

083165-1G 083165-10G
102839-078EA 72.31 USD
102839-078 102839-080
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride ≥95%
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride

Hazard: Irritant

Formula: C₁₁H₆Cl₃NO₂
MDL Number: MFCD00052556
CAS Number: 4462-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR