Isoxazole-5-carbonyl chloride ≥95%

Supplier: Matrix Scientific

083164-1G 083164-10G
102839-074EA 143.08 USD
102839-074 102839-076
Isoxazole-5-carbonyl chloride ≥95%
Isoxazole-5-carbonyl chloride

Hazard: Irritant

Formula: C₄H₂ClNO₂
MW: 131.52 g/mol
MDL Number: MFCD00067868
CAS Number: 62348-13-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR