Isonicotinoyl chloride hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
Pyridine-4-carbonyl Chloride Hydrochloride, 4-Pyridinoyl chloride hydrochloride
083136-1G
083136-10G
083136-50G
102839-010EA
72.31
USD
102839-010
102839-012
102839-014
Isonicotinoyl chloride hydrochloride ≥95%
Isonicotinoyl chloride hydrochloride
Hazard: Irritant
Formula:
C₆H₄ClNO MW: 178.01 g/mol |
MDL Number:
MFCD00012830 CAS Number: 39178-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...