2,3,5,6-Tetrachloroisonicotinoyl chloride ≥95%

Supplier: Matrix Scientific

083134-1G 083134-10G
102839-004EA 340 USD
102839-004 102839-006
2,3,5,6-Tetrachloroisonicotinoyl chloride ≥95%
2,3,5,6-Tetrachloroisonicotinoyl chloride

Hazard: Irritant

Formula: C₆Cl₅NO
MDL Number: MFCD08235235
CAS Number: 42935-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR