6-Chloronicotinoyl chloride ≥97%

Supplier: Matrix Scientific

083133-1G 083133-10G 083133-50G
102838-998EA 86.15 USD
102838-998 102839-000 102839-002
6-Chloronicotinoyl chloride ≥97%
6-Chloronicotinoyl chloride

Hazard: Irritant

Formula: C₆H₃Cl₂NO
MDL Number: MFCD00051775
CAS Number: 58757-38-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR