2-Chloro-6-methylpyridine-4-carbonyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-methylisonicotinoyl chloride

083132-1G 083132-10G
102838-994EA 260 USD
102838-994 102838-996
2-Chloro-6-methylpyridine-4-carbonyl chloride ≥95%
2-Chloro-6-methylpyridine-4-carbonyl chloride

Hazard: Irritant

Formula: C₇H₅Cl₂NO
MDL Number: MFCD00052829
CAS Number: 26413-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR