4-Chloropyridine-2-carbonyl chloride hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloropyridine-2-carbonyl chloride

083131-1G 083131-10G
102838-990EA 283.08 USD
102838-990 102838-992
4-Chloropyridine-2-carbonyl chloride hydrochloride ≥95%
4-Chloropyridine-2-carbonyl chloride hydrochloride

Hazard: Corrosive

Formula: C₆H₄Cl₃NO
MW: 212.46 g/mol
Melting Pt: 46…48 °C
MDL Number: MFCD08059127
CAS Number: 53750-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR