4-Propylbenzoyl chloride 95

Supplier: Matrix Scientific

Synonyms: 4-N-propylbenzoylchloride

083128-1G 083128-10G
102838-980EA 72.31 USD
102838-980 102838-982
4-Propylbenzoyl chloride 95
4-Propylbenzoyl chloride

Hazard: Irritant

Formula: C₁₀H₁₁ClO
MW: 182.65
Boiling Pt: 125
Density: 1.09
MDL Number: MFCD00000698
CAS Number: 52710-27-7
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR