2-Methyl-3-(trifluoromethyl)benzoyl chloride ≥97%

Supplier: Matrix Scientific

083123-1G 083123-10G
102838-968EA 241.54 USD
102838-968 102838-970
2-Methyl-3-(trifluoromethyl)benzoyl chloride ≥97%
2-Methyl-3-(trifluoromethyl)benzoyl chloride

Hazard: Irritant

Formula: C₉H₆ClF₃O
MDL Number: MFCD01631598
CAS Number: 261952-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR