2,6-Dimethylbenzoyl chloride ≥95%

Supplier: Matrix Scientific

083084-1G 083084-10G
102838-876EA 176.92 USD
102838-876 102838-878
2,6-Dimethylbenzoyl chloride ≥95%
2,6-Dimethylbenzoyl chloride

Hazard: Corrosive

Formula: C₉H₉ClO
MW: 168.62 g/mol
MDL Number: MFCD00052854
CAS Number: 21900-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR