2,3-Dimethylbenzoyl chloride ≥95%

Supplier: Matrix Scientific

083083-1G 083083-10G
102838-872EA 67.69 USD
102838-872 102838-874
2,3-Dimethylbenzoyl chloride ≥95%
2,3-Dimethylbenzoyl chloride

Hazard: Corrosive

Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 137°C/24 Torr
Melting Pt: 2°C
MDL Number: MFCD00045217
CAS Number: 21900-46-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR