2-[(5-Nitropyridin-2-yl)oxy]ethanol ≥97%

Supplier: Matrix Scientific

085065-1G 085065-10G
102844-154EA 156.92 USD
102844-154 102844-156
2-[(5-Nitropyridin-2-yl)oxy]ethanol ≥97%
2-[(5-Nitropyridin-2-yl)oxy]ethanol

Hazard: Irritant

Formula: C₇H₈N₂O₄
MDL Number: MFCD00052642
CAS Number: 143071-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR