1-Nitro-4-(2-pyridylthio)benzene ≥95%
Supplier: Matrix Scientific
Synonyms:
2-[(4-Nitrophenyl)sulphanyl]pyridine
085055-1G
085055-10G
102844-132EA
353.85
USD
102844-132
102844-134
1-Nitro-4-(2-pyridylthio)benzene ≥95%
1-Nitro-4-(2-pyridylthio)benzene
Hazard: Irritant
Formula:
C₁₁H₈N₂O₂S MW: 232.26 g/mol |
MDL Number:
MFCD00170204 CAS Number: 73322-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...